Assay Details
Target Gene Details
Entrez Gene ID: | 435921 |
Gene Name: | C-type lectin domain family 2, member f |
Gene Aliases: |
Clec2f-ps, Clr-c, Clrc, Clrz |
Location: |
Chr.6:129008803-129021342 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec2f | NM_001277202.1 | NP_001264131.1 | ||
| HQ875060.1 | ||||
| JF792240.1 |

Genomic Map