Assay Details
Target Gene Details
Entrez Gene ID: | 16653 |
Gene Name: | v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog |
Gene Aliases: |
AI929937, K-ras, Ki-ras, Kras-2, Kras2, p21B, ras |
Location: |
Chr.6:145216699-145250300 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kras | NM_021284.6 | 4 | 581 | NP_067259.4 |
| AK155064.1 | 4 | 582 | BAE33023.1 | |
| AK164065.1 | 4 | 582 | BAE37609.1 | |
| AY487807.1 | 3 | 405 | AAS58056.1 | |
| BC004642.1 | 4 | 569 | AAH04642.1 | |
| BC010202.1 | 4 | 569 | AAH10202.1 | |
| BC119495.1 | 5 | 445 | ||
| BC141051.1 | 3 | 407 | ||
| X00485.1 | 1 | 65 | CAA25160.1 |
More Information
Set Membership: |
|

Genomic Map