Assay Details
Target Gene Details
Entrez Gene ID: | 16653 |
Gene Name: | v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog |
Gene Aliases: |
AI929937, K-ras, Ki-ras, Kras-2, Kras2, p21B, ras |
Location: |
Chr.6:145216699-145250300 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kras | NM_021284.6 | NP_067259.4 | ||
| AK155064.1 | BAE33023.1 | |||
| AK164065.1 | BAE37609.1 | |||
| AY487807.1 | AAS58056.1 | |||
| BC004642.1 | AAH04642.1 | |||
| BC010202.1 | AAH10202.1 | |||
| BC119495.1 | ||||
| BC141051.1 |
More Information
Set Membership: |
|

Genomic Map