Assay Details
Target Gene Details
Entrez Gene ID: | 230376 |
Gene Name: | HAUS augmin-like complex, subunit 6 |
Gene Aliases: |
6230416J20Rik, 9830144E06, D4Ertd27e, mKIAA1574 |
Location: |
Chr.4:86578855-86612076 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Haus6 | NM_173400.2 | NP_775576.2 | ||
| AK040101.1 | BAC30511.1 | |||
| AK051016.1 | BAC34497.1 | |||
| AK077681.1 | BAC36953.1 | |||
| AK169222.1 | ||||
| AK173206.1 | BAD32484.1 | |||
| BC068237.1 | AAH68237.1 |

Genomic Map