Assay Details
Target Gene Details
Entrez Gene ID: | 77632 |
Gene Name: | PRAME family member 12 |
Gene Aliases: |
4930569K13Rik, AU023001, C86578 |
Location: |
Chr.4:144391674-144408467 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pramef12 | NM_029948.2 | NP_084224.1 | ||
| AK019795.1 | BAB31855.1 | |||
| AK167683.1 | ||||
| BC066168.1 | AAH66168.1 |
More Information
Set Membership: |
|

Genomic Map