Assay Details
Target Gene Details
Entrez Gene ID: | 433702 |
Gene Name: | nuclear cap binding protein subunit 1 |
Gene Aliases: |
AU014645, AW538051, CBP80 |
Location: |
Chr.4:46138511-46172403 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncbp1 | NM_001033201.3 | NP_001028373.2 | ||
| AK133991.1 | ||||
| AK134334.1 | BAE22102.1 | |||
| BC055777.1 | AAH55777.1 | |||
| BC138898.1 |
More Information
Set Membership: |
|

Genomic Map