Assay Details
Target Gene Details
Entrez Gene ID: | 246730 |
Gene Name: | 2'-5' oligoadenylate synthetase 1A |
Gene Aliases: |
L3 |
Location: |
Chr.5:120896256-120907543 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oas1a | NM_145211.2 | NP_660212.2 | ||
| AK149613.1 | BAE28990.1 | |||
| BC013715.1 | AAH13715.1 | |||
| BC057878.1 | AAH57878.1 | |||
| X04958.1 | CAA28620.1 |
More Information
Set Membership: |
|

Genomic Map