Assay Details
Target Gene Details
Entrez Gene ID: | 238722 |
Gene Name: | zinc finger protein 72 |
Gene Aliases: |
Zfp74 |
Location: |
Chr.13:74368682-74390861 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp72 | NM_001081680.1 | NP_001075149.1 | ||
| BC118036.1 | ||||
| BC118046.1 |
More Information
Set Membership: |
|

Genomic Map