Assay Details
Target Gene Details
Entrez Gene ID: | 211286 |
Gene Name: | ceroid-lipofuscinosis, neuronal 5 |
Gene Aliases: |
A730075N08Rik |
Location: |
Chr.14:103070216-103077630 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cln5 | NM_001033242.1 | 1 | 142 | NP_001028414.1 |
| AK032293.1 | BAC27797.1 | |||
| AK043247.1 | 1 | 66 | ||
| AK085741.1 | 1 | 143 | ||
| AK144635.1 | BAE25980.1 | |||
| BC025487.1 | AAH25487.1 | |||
| BC141314.1 | ||||
| BC141315.1 |

Genomic Map