Assay Details
Target Gene Details
Entrez Gene ID: | 319153 |
Gene Name: | histone cluster 1, H3i |
Gene Aliases: |
H3-H |
Location: |
Chr.13:21782915-21783397 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hist1h3i | NM_178207.2 | 1 | 409 | NP_835514.1 |
| BC107287.2 | AAI07288.1 | |||
| BC116383.2 | ||||
| BC127148.1 |
More Information
Set Membership: |
|

Genomic Map