Assay Details
Target Gene Details
Entrez Gene ID: | 238662 |
Gene Name: | spermatogenesis associated 31 subfamily D, member 1B |
Gene Aliases: |
EG238662, Fam75d1b, Gm4934 |
Location: |
Chr.13:59712284-59719289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata31d1b | NM_001167593.1 | NP_001161065.1 | ||
| AK133300.1 |
More Information
Set Membership: |
|

Genomic Map