Assay Details
Target Gene Details
Entrez Gene ID: | 97863 |
Gene Name: | expressed sequence C78339 |
Gene Aliases: |
- |
Location: |
Chr.13:46669443-46678064 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C78339 | NM_001033192.2 | NP_001028364.1 | ||
| AK090371.1 | ||||
| AK141282.1 | BAE24634.1 | |||
| AK171039.1 | BAE42204.1 | |||
| AK218359.1 |

Genomic Map