Assay Details
Target Gene Details
Entrez Gene ID: | 12445 |
Gene Name: | cyclin D3 |
Gene Aliases: |
9230106B05Rik, AA682053, AL024085, AW146355, C78795 |
Location: |
Chr.17:47504738-47599689 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnd3 | NM_001081635.1 | NP_001075104.1 | ||
| NM_001081636.1 | NP_001075105.1 | |||
| NM_007632.2 | NP_031658.1 | |||
| AK083384.1 | ||||
| AK159503.1 | BAE35136.1 | |||
| AK161758.1 | BAE36561.1 | |||
| AK165191.1 | ||||
| AK168290.1 | BAE40235.1 | |||
| AK213147.1 | ||||
| BC004076.1 | AAH04076.1 | |||
| BC005605.1 | AAH05605.1 | |||
| M86183.1 | AAA37504.1 |
More Information
Set Membership: |
|

Genomic Map