Assay Details
Target Gene Details
Entrez Gene ID: | 75202 |
Gene Name: | sperm acrosome associated 6 |
Gene Aliases: |
4930546H06Rik, 9530079D04Rik, B230206P06Rik, Ncrna00085, let-7e, mir-125a, mir-99b |
Location: |
Chr.17:17824821-17843010 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spaca6 | NM_001162909.1 | NP_001156381.1 | ||
| AK016052.1 | BAB30092.1 | |||
| AK080293.1 | BAC37867.1 | |||
| BC171982.1 | ||||
| BC171984.1 |

Genomic Map