Assay Details
Target Gene Details
Entrez Gene ID: | 69083 |
Gene Name: | sulfotransferase family, cytosolic, 1C, member 2 |
Gene Aliases: |
1810008N17Rik, ST1C1, ST1C2, Sult1c1 |
Location: |
Chr.17:53829637-53845958 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sult1c2 | NM_026935.4 | NP_081211.3 | ||
| AK007384.1 | BAB25002.1 | |||
| AK144336.1 | BAE25838.1 | |||
| AY005469.1 | AAG00823.1 | |||
| BC022665.1 | AAH22665.1 |
More Information
Set Membership: |
|

Genomic Map