Assay Details
Target Gene Details
Entrez Gene ID: | 17158 |
Gene Name: | mannosidase 2, alpha 1 |
Gene Aliases: |
Mana-2, Mana2, Map-2 |
Location: |
Chr.17:64601649-64755110 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Man2a1 | NM_008549.2 | 17 | 2698 | NP_032575.2 |
| BC138371.1 | 17 | 2683 | ||
| BC138372.1 | 17 | 2683 | ||
| X61172.1 | 17 | 2698 | CAA43480.1 |

Genomic Map