Assay Details
Target Gene Details
Entrez Gene ID: | 100502766 |
Gene Name: | kinesin family member C1 |
Gene Aliases: |
100042970, Gm4137, HSET, KNSL2, Kifc5a, Knsl2a, Tctex-7, Tctex-7A, Tctex7, Tctex7a |
Location: |
Chr.17:33875664-33890633 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kifc1 | NM_001195298.1 | NP_001182227.1 | ||
| AF221102.1 | AAF34646.1 | |||
| AI326799.1 | ||||
| AK133900.1 | ||||
| BC003753.1 | AAH03753.1 | |||
| BC057162.1 | AAH57162.1 | |||
| BC100328.1 | AAI00329.1 | |||
| D49544.1 | BAA19676.1 |
More Information
Set Membership: |
|

Genomic Map