Assay Details
Target Gene Details
Entrez Gene ID: | 68774 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 6D |
Gene Aliases: |
1110058E16Rik, AI447446, Ms4a11 |
Location: |
Chr.19:11586606-11604804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a6d | NM_026835.2 | NP_081111.1 | ||
| AB026047.1 | BAB18759.1 | |||
| AF237911.1 | AAK37593.1 | |||
| AK004295.1 | BAB23253.1 | |||
| AK152859.1 | ||||
| AK156878.1 | ||||
| AY039037.1 | AAK92697.1 | |||
| BC018331.1 | AAH18331.1 | |||
| BC029738.1 | AAH29738.1 | |||
| BG087418.2 |
More Information
Set Membership: |
|

Genomic Map