Assay Details
Target Gene Details
Entrez Gene ID: | 338360 |
Gene Name: | RIKEN cDNA B430212C06 gene |
Gene Aliases: |
- |
Location: |
Chr.18:67321209-67344103 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B430212C06Rik | NR_033214.1 | |||
| AK046629.1 | 2 | 898 | BAC32813.1 | |
| AK087569.1 | ||||
| AK148585.1 | ||||
| AK164742.1 |

Genomic Map