Assay Details
Target Gene Details
Entrez Gene ID: | 214812 |
Gene Name: | zinc finger protein 609 |
Gene Aliases: |
9830165N24, BC016271, Znf609, mKIAA0295 |
Location: |
Chr.9:65691578-65827564 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp609 | NM_172536.3 | NP_766124.2 | ||
| AK036715.1 | BAC29549.1 | |||
| BC016271.1 | ||||
| BC058943.1 | AAH58943.1 | |||
| BC150683.1 |

Genomic Map