Assay Details
Target Gene Details
Entrez Gene ID: | 12442 |
Gene Name: | cyclin B2 |
Gene Aliases: |
CycB2 |
Location: |
Chr.9:70407689-70421554 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb2 | NM_007630.2 | 9 | 1231 | NP_031656.2 |
| AA033463.1 | 2 | 120 | ||
| AK013312.1 | 9 | 1200 | BAB28785.1 | |
| AK076122.1 | 9 | 1225 | BAC36200.1 | |
| BC008247.1 | 9 | 1147 | AAH08247.1 | |
| CT010286.1 | 9 | 1117 | CAJ18494.1 | |
| X66032.1 | 9 | 1218 | CAA46831.1 |
More Information
Set Membership: |
|

Genomic Map