Assay Details
Target Gene Details
Entrez Gene ID: | 67618 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase |
Gene Aliases: |
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5 |
Location: |
Chr.9:4294793-4309509 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aasdhppt | NM_026276.3 | 3 | 559 | NP_080552.3 |
| AK007737.1 | 3 | 560 | BAB25224.1 | |
| AK008554.1 | BAB25740.1 | |||
| AK010557.1 | 3 | 519 | BAB27026.1 | |
| AK013111.1 | 3 | 513 | BAB28656.1 | |
| AK014188.1 | 3 | 502 | ||
| AK018191.1 | 2 | 205 | BAB31116.1 | |
| AK028700.1 | 3 | 516 | ||
| AK029523.1 | 3 | 501 | ||
| AK031766.1 | ||||
| AK076099.1 | 3 | 514 | BAC36182.1 | |
| AK076866.1 | 3 | 533 | BAC36512.1 | |
| AK081818.1 | 3 | 535 | ||
| AK088977.1 | ||||
| AK144671.1 | 3 | 533 | BAE26000.1 | |
| AK214035.1 | ||||
| AK215948.1 | ||||
| BC030043.1 | 3 | 494 | ||
| BC038013.1 | 3 | 530 | AAH38013.1 | |
| BC049851.1 | 3 | 520 | AAH49851.1 |
More Information
Set Membership: |
|

Genomic Map