Assay Details
Target Gene Details
Entrez Gene ID: | 67618 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase |
Gene Aliases: |
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5 |
Location: |
Chr.9:4294793-4309509 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aasdhppt | NM_026276.3 | 2 | 337 | NP_080552.3 |
| AK007737.1 | 2 | 338 | BAB25224.1 | |
| AK008554.1 | 2 | 302 | BAB25740.1 | |
| AK010557.1 | 2 | 297 | BAB27026.1 | |
| AK013111.1 | 2 | 291 | BAB28656.1 | |
| AK014188.1 | 2 | 280 | ||
| AK018191.1 | BAB31116.1 | |||
| AK028700.1 | 2 | 294 | ||
| AK029523.1 | 2 | 279 | ||
| AK031766.1 | 2 | 281 | ||
| AK076099.1 | 2 | 292 | BAC36182.1 | |
| AK076866.1 | 2 | 311 | BAC36512.1 | |
| AK081818.1 | 2 | 313 | ||
| AK088977.1 | 2 | 277 | ||
| AK144671.1 | 2 | 311 | BAE26000.1 | |
| AK188246.1 | ||||
| AK214035.1 | ||||
| AK215948.1 | ||||
| BC030043.1 | 2 | 272 | ||
| BC038013.1 | 2 | 308 | AAH38013.1 | |
| BC049851.1 | 2 | 298 | AAH49851.1 |
More Information
Set Membership: |
|

Genomic Map