Assay Details
Target Gene Details
Entrez Gene ID: | 26611 |
Gene Name: | reticulocalbin 2 |
Gene Aliases: |
AA408742, Tcbp49 |
Location: |
Chr.9:56041845-56059083 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcn2 | NM_001278274.1 | 1 | 156 | NP_001265203.1 |
| NM_011992.2 | 1 | 156 | NP_036122.2 | |
| AF049125.1 | 1 | 141 | AAC05132.1 | |
| AK076153.1 | ||||
| AK077486.1 | 1 | 132 | BAC36825.1 | |
| AK077734.1 | 1 | 132 | BAC36986.1 | |
| AK078106.1 | 1 | 127 | ||
| AK079156.1 | 1 | 132 | ||
| AK169765.1 | 1 | 130 | BAE41353.1 | |
| BB638743.1 | 1 | 157 | ||
| BC132320.1 | 1 | 121 | ||
| BC145668.1 | 1 | 121 |
More Information
Set Membership: |
|

Genomic Map