Assay Details
Target Gene Details
Entrez Gene ID: | 382053 |
Gene Name: | carboxylesterase 3A |
Gene Aliases: |
Ces3, Ces31, Es31 |
Location: |
Chr.8:105048577-105061928 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces3a | NM_001164681.1 | NP_001158153.1 | ||
| NM_198672.1 | NP_941074.1 | |||
| AK138932.1 | BAE23821.1 | |||
| BC048380.1 | AAH48380.1 | |||
| BC057187.1 | AAH57187.1 | |||
| BC057188.1 | AAH57188.1 | |||
| BC061004.1 | ||||
| CB952677.1 | ||||
| L11333.1 | ||||
| S64130.1 | AAB27606.1 |
More Information
Set Membership: |
|

Genomic Map