Assay Details
Target Gene Details
Entrez Gene ID: | 22768 |
Gene Name: | zinc finger protein 2, Y linked |
Gene Aliases: |
Zfy-2 |
Location: |
Chr.Y:2106015-2170409 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfy2 | NM_009571.2 | 1 | 192 | NP_033597.2 |
| AK133222.1 | 1 | 193 | BAE21566.1 | |
| M24401.1 | 1 | 153 | AAA56845.1 |
More Information
Set Membership: |
|

Genomic Map