Assay Details
Target Gene Details
Entrez Gene ID: | 11571 |
Gene Name: | cysteine-rich secretory protein 1 |
Gene Aliases: |
AW324342, Aeg1, CRISP-1 |
Location: |
Chr.17:40293758-40319246 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crisp1 | NM_009638.3 | 8 | 766 | NP_033768.3 |
| AK009449.1 | 8 | 767 | BAB26295.1 | |
| BC011150.1 | 8 | 749 | AAH11150.1 | |
| L05559.1 | 8 | 736 | AAA37460.1 | |
| M92849.1 | 8 | 732 | AAA37185.1 |

Genomic Map