Assay Details
Target Gene Details
Entrez Gene ID: | 16688 |
Gene Name: | keratin 6B |
Gene Aliases: |
BB144589, Krt2-6b, mK6[b] |
Location: |
Chr.15:101676023-101680289 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt6b | NM_010669.2 | 1 | 64 | NP_034799.2 |
| AK137688.1 | 1 | 63 | BAE23462.1 | |
| BB565369.1 | 1 | 64 | ||
| BC122884.1 | ||||
| BC124556.1 |

Genomic Map