Assay Details
Target Gene Details
Entrez Gene ID: | 71138 |
Gene Name: | transmembrane protein 217 |
Gene Aliases: |
4933413N12Rik, EG622644 |
Location: |
Chr.17:29519143-29549689 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem217 | NM_001162901.1 | 3 | 363 | NP_001156373.1 |
| NM_001162902.1 | 2 | 271 | NP_001156374.1 | |
| AK016810.1 | 3 | 364 | ||
| BC116865.1 | 2 | 235 | ||
| BC116867.1 | 3 | 327 | ||
| BC144829.1 | 2 | 235 |

Genomic Map