Assay Details
Target Gene Details
Entrez Gene ID: | 81845 |
Gene Name: | G patch domain and ankyrin repeats 1 |
Gene Aliases: |
Bat-4, Bat4, D17H6S54E, G5 |
Location: |
Chr.17:35121473-35124815 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpank1 | NM_001128597.1 | NP_001122069.1 | ||
| NM_032460.2 | 1 | 72 | NP_115849.2 | |
| AB221568.1 | 1 | 38 | ||
| AK028189.1 | BAC25800.1 | |||
| BC057902.1 | AAH57902.1 | |||
| BC120637.1 | 1 | 73 | ||
| BC120639.1 | 1 | 73 | ||
| CJ069410.1 |

Genomic Map