Assay Details
Target Gene Details
Entrez Gene ID: | 246729 |
Gene Name: | 2'-5' oligoadenylate synthetase 1H |
Gene Aliases: |
- |
Location: |
Chr.5:120861422-120873506 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oas1h | NM_001159934.1 | 4 | 1235 | NP_001153406.1 |
| NM_145228.2 | 6 | 1625 | NP_660263.1 | |
| AB067530.1 | 6 | 1625 | BAB84130.1 | |
| AK135899.1 | 4 | 1240 |
More Information
Set Membership: |
|

Genomic Map