Assay Details
Target Gene Details
Entrez Gene ID: | 387356 |
Gene Name: | taste receptor, type 2, member 131 |
Gene Aliases: |
T2R31, Tas2r31, mGR31, mt2r61 |
Location: |
Chr.6:132956912-132957844 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r131 | NM_207030.1 | 1 | 882 | NP_996913.1 |
| BC145798.1 | 1 | 957 |
More Information
Set Membership: |
|

Genomic Map