Assay Details
Target Gene Details
Entrez Gene ID: | 108096 |
Gene Name: | solute carrier organic anion transporter family, member 1a5 |
Gene Aliases: |
Oatp-3, Oatp3, Slc21a7 |
Location: |
Chr.6:142234227-142322981 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slco1a5 | NM_001267707.1 | 15 | 2262 | NP_001254636.1 |
| NM_130861.3 | 15 | 2210 | NP_570931.1 | |
| AF240694.1 | 15 | 2180 | AAK39416.1 | |
| AK029706.1 | 15 | 2212 | ||
| BC013594.1 | 15 | 2224 | AAH13594.1 |
More Information
Set Membership: |
|

Genomic Map