Assay Details
Target Gene Details
Entrez Gene ID: | 233871 |
Gene Name: | ataxin 2-like |
Gene Aliases: |
A2D, A2LG, A2RP, A2lp, Atxnl2 |
Location: |
Chr.7:126491708-126503644 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atxn2l | NM_183020.1 | 23 | 3352 | NP_898841.1 |
| AK155062.1 | 22 | 3995 | ||
| AK158626.1 | 23 | 3564 | BAE34586.1 | |
| AK168745.1 | 22 | 3665 | BAE40586.1 | |
| BC034083.1 | 17 | 2431 | AAH34083.1 | |
| BC043451.1 | 17 | 3106 | AAH43451.1 | |
| BC054483.1 | 23 | 3352 | AAH54483.1 |
More Information
Set Membership: |
|

Genomic Map