Assay Details
Target Gene Details
Entrez Gene ID: | 231290 |
Gene Name: | solute carrier family 10 (sodium/bile acid cotransporter family), member 4 |
Gene Aliases: |
E130304D01 |
Location: |
Chr.5:73006904-73012955 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc10a4 | NM_173403.2 | 1 | 276 | NP_775579.2 |
| AK087479.1 | 1 | 277 | BAC39890.1 | |
| BC132529.1 | 1 | 188 | ||
| BC138048.1 | 1 | 188 |
More Information
Set Membership: |
|

Genomic Map