Assay Details
Target Gene Details
Entrez Gene ID: | 625253 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 9 pseudogene |
Gene Aliases: |
EG625253 |
Location: |
Chr.X:157540804-157541750 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6568 | NR_040420.2 | 1 | 307 | |
| BB810489.1 | 1 | 191 |

Genomic Map