Assay Details
Target Gene Details
Entrez Gene ID: | 67263 |
Gene Name: | zinc finger SWIM-type containing 6 |
Gene Aliases: |
2900036G02Rik, mKIAA1577 |
Location: |
Chr.13:107724617-107890064 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zswim6 | NM_145456.2 | 1 | 261 | NP_663431.2 |
| CJ181852.1 | 2 | 229 |

Genomic Map