Assay Details
Target Gene Details
Entrez Gene ID: | 56517 |
Gene Name: | solute carrier family 22 (organic cation transporter), member 21 |
Gene Aliases: |
Octn3, Slc22a9 |
Location: |
Chr.11:53950187-53980818 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a21 | NM_019723.2 | 1 | 49 | NP_062697.1 |
| AB018436.1 | 1 | 49 | BAA78343.1 | |
| AK080177.1 | 1 | 356 | ||
| AK133431.1 | BAE21654.1 | |||
| AK144849.1 | 1 | 390 | ||
| BC132147.1 | ||||
| BC137908.1 |
More Information
Set Membership: |
|

Genomic Map