Assay Details
Target Gene Details
Entrez Gene ID: | 67019 |
Gene Name: | ARP6 actin-related protein 6 |
Gene Aliases: |
2010200J04Rik, AU044443, Arp6, ArpX, CDA12 |
Location: |
Chr.10:89711966-89732295 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr6 | NM_025914.2 | 11 | 1194 | NP_080190.1 |
| AK008409.1 | 11 | 1195 | BAB25654.1 | |
| AK088571.1 | 11 | 1180 | BAC40429.1 | |
| BC062137.1 | 11 | 1171 | AAH62137.1 |
More Information
Set Membership: |
|

Genomic Map