Assay Details
Target Gene Details
Entrez Gene ID: | 259040 |
Gene Name: | olfactory receptor 1416 |
Gene Aliases: |
MOR103-3 |
Location: |
Chr.1:92479681-92480619 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr1416 | NM_147038.1 | 1 | 897 | NP_667249.1 |
| BC104078.1 | 1 | 896 | AAI04079.1 | |
| BC104079.2 | 1 | 896 | AAI04080.1 |
More Information
Set Membership: |
|

Genomic Map