Assay Details
Target Gene Details
Entrez Gene ID: | 259041 |
Gene Name: | olfactory receptor 1414 |
Gene Aliases: |
MOR103-2 |
Location: |
Chr.1:92511048-92518515 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr1414 | NM_147039.2 | 2 | 438 | NP_667250.1 |
| BC120830.1 | 1 | 265 |
More Information
Set Membership: |
|

Genomic Map