Assay Details
Target Gene Details
Entrez Gene ID: | 241943 |
Gene Name: | coiled-coil domain containing 144B |
Gene Aliases: |
- |
Location: |
Chr.3:36007247-36053547 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc144b | NM_178418.4 | 12 | 1831 | NP_848505.2 |
| BC050811.1 | 12 | 1833 | AAH50811.1 |
Target Gene Details
Entrez Gene ID: | 654424 |
Gene Name: | methylcrotonoyl-Coenzyme A carboxylase 1 (alpha), opposite strand |
Gene Aliases: |
A330050B17Rik |
Location: |
Chr.3:35999913-36008828 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mccc1os | NR_029456.1 | 3 | 3517 | |
| AK138522.1 | 3 | 3519 | BAE23692.1 |

Genomic Map