Assay Details
Target Gene Details
Entrez Gene ID: | 20439 |
Gene Name: | seven in absentia 2 |
Gene Aliases: |
AA415433, Sinh2 |
Location: |
Chr.3:58674949-58692388 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Siah2 | NM_009174.3 | 1 | 90 | NP_033200.2 |
| AK139626.1 | 1 | 66 | BAE24087.1 | |
| AK149316.1 | 1 | 104 | BAE28809.1 | |
| BC058400.1 | 1 | 90 | AAH58400.1 |
More Information
Set Membership: |
|

Genomic Map