Assay Details
Target Gene Details
Entrez Gene ID: | 52477 |
Gene Name: | angel homolog 2 (Drosophila) |
Gene Aliases: |
2610307I21Rik, 5730410O10Rik, AI845692, D1Ertd396e, D1Ertd654e |
Location: |
Chr.1:190925108-190946962 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angel2 | NM_001199020.1 | 10 | 1825 | NP_001185949.1 |
| NM_021421.4 | 9 | 1960 | NP_067396.3 | |
| NR_037578.1 | 9 | 1919 | ||
| NR_037579.1 | 8 | 1634 | ||
| AB041602.1 | 7 | 1493 | BAA95085.1 | |
| AK077039.1 | 9 | 1907 | ||
| AK088834.1 | 8 | 1593 | BAC40602.1 | |
| AK135709.1 | 10 | 1822 | BAE22620.1 | |
| AK158731.1 | 9 | 1976 | BAE34634.1 | |
| AK163397.1 | ||||
| AK166619.1 | 10 | 1824 | BAE38898.1 | |
| AK169653.1 | 8 | 1544 | BAE41277.1 | |
| AK203912.1 | 1 | 241 | ||
| AK217899.1 | 1 | 247 | ||
| BC024907.1 | 9 | 1920 | AAH24907.1 | |
| BF181986.1 |
More Information
Set Membership: |
|

Genomic Map