Assay Details
Target Gene Details
Entrez Gene ID: | 14201 |
Gene Name: | four and a half LIM domains 3 |
Gene Aliases: |
SLIM2 |
Location: |
Chr.4:124695963-124708611 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhl3 | NM_010213.3 | 6 | 1360 | NP_034343.2 |
More Information
Set Membership: |
|

Genomic Map