Assay Details
Target Gene Details
Entrez Gene ID: | 245607 |
Gene Name: | G protein-coupled receptor associated sorting protein 2 |
Gene Aliases: |
5330440H13Rik, GASP-2, GASP2, Prpl5 |
Location: |
Chr.X:135839034-135844730 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprasp2 | NM_001163015.1 | NP_001156487.1 | ||
| NM_001163016.1 | NP_001156488.1 | |||
| NM_001163017.1 | NP_001156489.1 | |||
| AK030638.1 | ||||
| AK081764.1 | BAC38324.1 | |||
| BC117923.2 | ||||
| BC117924.2 |
More Information
Set Membership: |
|

Genomic Map