Assay Details
Target Gene Details
Entrez Gene ID: | 100039801 |
Gene Name: | claudin 24 |
Gene Aliases: |
ENSMUSG00000061974, Gm10107 |
Location: |
Chr.8:47822143-47822805 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cldn24 | NM_001111318.1 | 1 | 54 | NP_001104788.1 |
More Information
Set Membership: |
|

Genomic Map