Assay Details
Target Gene Details
Entrez Gene ID: | 14082 |
Gene Name: | Fas (TNFRSF6)-associated via death domain |
Gene Aliases: |
Mort1/FADD |
Location: |
Chr.7:144578323-144582436 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fadd | NM_010175.5 | NP_034305.1 | ||
| AK007648.1 | ||||
| AK031396.1 | BAC27384.1 | |||
| AK084808.1 | BAC39283.1 | |||
| AK169798.1 | BAE41374.1 | |||
| AK170927.1 | BAE42120.1 | |||
| BC004584.1 | AAH04584.1 | |||
| BC021400.1 | AAH21400.1 | |||
| U43184.1 | AAA97876.1 | |||
| U50406.1 | AAB07789.1 |
More Information
Set Membership: |
|

Genomic Map