Assay Details
Target Gene Details
Entrez Gene ID: | 18416 |
Gene Name: | ornithine transcarbamylase |
Gene Aliases: |
AI265390, Sf, spf |
Location: |
Chr.X:10252297-10321024 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otc | NM_008769.3 | NP_032795.1 | ||
| AK050930.1 | BAC34465.1 | |||
| BC024893.1 | AAH24893.1 | |||
| M17030.1 | AAA39865.1 |
More Information
Set Membership: |
|

Genomic Map