Assay Details
Target Gene Details
Entrez Gene ID: | 434234 |
Gene Name: | RIKEN cDNA 2610020H08 gene |
Gene Aliases: |
- |
Location: |
Chr.7:119794130-119882695 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610020H08Rik |
Target Gene Details
Entrez Gene ID: | 233805 |
Gene Name: | DCN1, defective in cullin neddylation 1, domain containing 3 (S. cerevisiae) |
Gene Aliases: |
1700020A13Rik, BC030335 |
Location: |
Chr.7:119853158-119896169 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcun1d3 | NM_001163703.1 | NP_001157175.1 | ||
| NM_173408.3 | NP_775584.1 | |||
| AK029043.1 | BAC26261.1 | |||
| AK155160.1 | BAE33084.1 | |||
| BC030335.1 | AAH30335.1 |
More Information
Set Membership: |
|

Genomic Map